Learn More
CIB1 Mouse anti-Human, Clone: OTI1D7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811149
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the calcium-binding protein family. The specific function of this protein has not yet been determined; however this protein is known to interact with DNA-dependent protein kinase and may play a role in kinase-phosphatase regulation of DNA end joining. This protein also interacts with integrin alpha(IIb)beta(3), which may implicate this protein as a regulatory molecule for alpha(IIb)beta(3).Specifications
| CIB1 | |
| Monoclonal | |
| Unconjugated | |
| CIB1 | |
| CIB, CIBP, KIP1, PRKDCIP, SIP2-28 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10519 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1D7 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q99828 | |
| CIB1 | |
| Human recombit protein fragment corresponding to amino acids 2-191 of human CIB1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.