missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIDEA Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98783C
This item is not returnable.
View return policy
Description
CIDEA Polyclonal antibody specifically detects CIDEA in Human samples. It is validated for Western Blot
Specifications
| CIDEA | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 650 | |
| cell death activator CIDE-A, cell death-inducing DFFA-like effector aCIDE-A | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CIDEA. | |
| 0.1 mL | |
| Apoptosis | |
| 1149 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction