missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIDEA Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76950F
This item is not returnable.
View return policy
Description
CIDEA Polyclonal antibody specifically detects CIDEA in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CIDEA | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| cell death activator CIDE-A, cell death-inducing DFFA-like effector aCIDE-A | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of mouse CIDE-A. The immunogen is located within the last 50 amino acids of CIDE-A. Amino Acid Squence: GDTEEQPSPKPSTKG | |
| 0.1 mL | |
| Apoptosis | |
| 1149 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction