missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIDEB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101329
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CIDEB.
CIDEB activates apoptosis.
Specifications
| CIDEB | |
| Polyclonal | |
| Unconjugated | |
| CIDEB | |
| 1110030C18Rik; AI790179; Cell death activator CIDE-B; cell death-inducing DFFA-like effector B; cell death-inducing DNA fragmentation factor, alpha subunit-like effector B; CID; Cideb; CIDE-B; DFFA-like B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12684, 27141 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70303, Q9UHD4 | |
| CIDEB | |
| A synthesized peptide derived from human CIDEB(Accession Q9UHD4), corresponding to amino acid residues T97-F147. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction