Learn More
Cidofovir Diphosphate Trisodium Salt (>85%), TRC
Specifications
Specifications
| Chemical Name or Material | Cidofovir Diphosphate Trisodium Salt |
| CAS | 352525-57-6 |
| Molecular Formula | C8 H13 N3 Na3 O12 P3 |
| Quantity | 10 mg |
| Synonym | Diphosphoric acid Monoanhydride with [[(1S)-2-(4-Amino-2-oxo-1(2H)-pyrimidinyl)-1-(hydroxymethyl)ethoxy]methyl]phosphonic Acid Trisodium Salt |
| InChI Formula | InChI=1S/C8H16N3O12P3.3Na/c9-7-1-2-11(8(13)10-7)3-6(4-12)21-5-24(14,15)22-26(19,20)23-25(16,17)18;;;/h1-2,6,12H,3-5H2,(H,14,15)(H,19,20)(H2,9,10,13)(H2,16,17,18);;;/q;3*+1/p-3/t6-;;;/m0.../s1 |
| SMILES | NC1=NC(=O)N(C[C@@H](CO)OCP(=O)(O[Na])OP(=O)(O[Na])OP(=O)(O)O[Na])C=C1 |
| IUPAC Name | [[(2S)-1-(4-amino-2-oxopyrimidin-1-yl)-3-hydroxypropan-2-yl]oxymethyl-[[hydroxy(sodiooxy)phosphoryl]oxy-sodiooxyphosphoryl]oxyphosphoryl]oxysodium |
| Molecular Weight (g/mol) | 505.092 |
| Formula Weight | 504.941 |