Learn More
Cilastatin, MedChemExpress
Product Code. 16488036
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488036 100 mg 100mg
16458308 10 mg 10mg
16424197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16488036 Supplier Medchem Express Supplier No. HYA0166
MedChemExpress Cilastatin (MK0791) is a reversible, competitive renal dehydropeptidase I inhibitor with an IC50 of 0.1 μM. Cilastatin inhibits the bacterial metallob-lactamase enzyme CphA with an IC50 of 178 μM. Cilastatin is an antibacterial adjunct.
Cilastatin (MK0791) is a reversible, competitive renal dehydropeptidase I inhibitor with an IC50 of 0.1 μM. Cilastatin inhibits the bacterial metallob-lactamase enzyme CphA with an IC50 of 178 μM. Cilastatin is an antibacterial adjunct.
Specifications
Chemical Name or Material Cilastatin
Physical Form Solid
CAS 82009-34-5
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C16H26N2O5S
For Use With (Application) COVID-19-immunoregulation
Synonym MK0791
Solubility Information DMSO : 100 mg/mL (278.98 mM; Need ultrasonic)
SMILES O=C(O)/C(NC([C@@H]1C(C)(C)C1)=O)=C/CCCCSC[C@H](N)C(O)=O
Molecular Weight (g/mol) 358.45
Formula Weight 358.45
Percent Purity 99.37%
Grade Research
Show More Show Less
Research purposes only