missing translation for 'onlineSavingsMsg'
Learn More

Cilengitide, MedChemExpress

Product Code. 16473047
Encompass
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473047

Brand: Medchem Express HY16141

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Cilengitide (EMD 121974) is a potent and selective inhibitor of the integrins ανβ3 and ανβ5. Cilengitide inhibits binding of isolated ανβ3 and ανβ5 to Vitronectin with an IC50 value of 4 and 79 nM, respectively .

Cilengitide (EMD 121974) is a potent and selective inhibitor of the integrins ανβ3 and ανβ5. Cilengitide inhibits binding of isolated ανβ3 and ανβ5 to Vitronectin with an IC50 value of 4 and 79 nM, respectively .

Specifications

Chemical Name or Material Cilengitide
Physical Form Solid
CAS 188968-51-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C27H40N8O7
For Use With (Application) Cancer-programmed cell death
Synonym EMD 121974
Solubility Information H2O : 100 mg/mL (169.88 mM; Need ultrasonic) ∣DMSO : ≥ 44 mg/mL (74.75 mM)
SMILES O=C(NCC(N[C@H](C(N[C@H](CC1=CC=CC=C1)C(N([C@H]2C(C)C)C)=O)=O)CC(O)=O)=O)[C@H](CCCNC(N)=N)NC2=O
Molecular Weight (g/mol) 588.66
Formula Weight 588.66
Percent Purity 95.18%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.