missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cinepazide Maleate, TRC
Specifications
Specifications
| Chemical Name or Material | Cinepazide Maleate |
| CAS | 26328-04-1 |
| Molecular Formula | C22H31N3O5 C4H4O4 |
| Quantity | 50 mg |
| Synonym | 1-[2-oxo-2-(1-pyrrolidinyl)ethyl]-4-[1-oxo-3-(3,4,5-trimethoxyphenyl)-2-propenyl]piperazine (2Z)-2-butenedioate (1:1), Brendil, MD 67350, NSC 291562, Vasodistal |
| InChI Formula | InChI=1S/C22H31N3O5.C4H4O4/c1-28-18-14-17(15-19(29-2)22(18)30-3)6-7-20(26)25-12-10-23(11-13-25)16-21(27)24-8-4-5-9-24;5-3(6)1-2-4(7)8/h6-7,14-15H,4-5,8-13,16H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | COc1cc(C=CC(=O)N2CCN(CC(=O)N3CCCC3)CC2)cc(OC)c1OC.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;1-[4-(2-oxo-2-pyrrolidin-1-ylethyl)piperazin-1-yl]-3-(3,4,5-trimethoxyphenyl)prop-2-en-1-one |
| Molecular Weight (g/mol) | 533.6 |
| Formula Weight | 533.2373 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?