missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cingulin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 463.00
Specifications
| Antigen | Cingulin |
|---|---|
| Dilution | Western Blot 1:200-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695531
|
Novus Biologicals
NBP2-92240-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677461
|
Novus Biologicals
NBP2-92240-0.1ml |
0.1 mL |
€ 463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cingulin Polyclonal antibody specifically detects Cingulin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Cingulin | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 57530 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| cingulin, DKFZp779N1112, KIAA1319 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 904-1203 of human CGN (NP_065821.1). TSGGLSRLQDEIQRLRQALQASQAERDTARLDKELLAQRLQGLEQEAENKKRSQDDRARQLKGLEEKVSRLETELDEEKNTVELLTDRVNRGRDQVDQLRTELMQERSARQDLECDKISLERQNKDLKTRLASSEGFQKPSASLSQLESQNQLLQERLQAEEREKTVLQSTNRKLERKVKELSIQIEDERQHVNDQKDQLSLRVKALKRQVDEAEEEIERLDGLRKKAQREVEEQHEVNEQLQARIKSLEKDSWRKASRSAAESALKNEGLSSDEEFDSVYDPSSIASLLTESNLQTSSC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title