missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CINP Antibody (1G10), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59486AF750
This item is not returnable.
View return policy
Description
CINP Monoclonal antibody specifically detects CINP in Human samples. It is validated for Western Blot, ELISA
Specifications
| CINP | |
| Monoclonal | |
| Alexa Fluor 750 | |
| CDK2-interacting protein, cyclin-dependent kinase 2 interacting protein, cyclin-dependent kinase 2-interacting protein, MGC849 | |
| Recombinant human CINP (1-212aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 51550 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 1G10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction