missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CINP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37914-20ul
This item is not returnable.
View return policy
Description
CINP Polyclonal antibody specifically detects CINP in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
| CINP | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| CDK2-interacting protein, cyclin-dependent kinase 2 interacting protein, cyclin-dependent kinase 2-interacting protein, MGC849 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-212 of human CINP (NP_116019.1).,, Sequence:, MEAKTLGTVTPRKPVLSVSARKIKDNAADWHNLILKWETLNDAGFTTANNIANLKISLLNKDKIELDSSSPASKENEEKVCLEYNEELEKLCEELQATLDGLTKIQVKMEKLSSTTKGICELENYHYGEESKRPPLFHTWPTTHFYEVSHKLLEMYRKELLLKRTVAKELAHTGDPDLTLSYLSMWLHQPYVESDSRLHLESMLLETGHRAL | |
| 20 μL | |
| Cell Cycle and Replication | |
| 51550 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction