missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIP2A Antibody (2G10), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-59722IR
This item is not returnable.
View return policy
Description
CIP2A Monoclonal specifically detects CIP2A in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| CIP2A | |
| Monoclonal | |
| DyLight 755 | |
| Q8TCG1 | |
| CIP2A | |
| A synthetic peptide made to a C-terminal portion of the human CIP2A protein. [UniProt# Q8TCG1] | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 57650 | |
| Human, Mouse, Rat, Primate | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| 2G10 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, KnockDown Validated | |
| Cancerous inhibitor of PP2A, CIP2A, FLJ12850, KIAA1524, MGC163436, p90, p90 autoantigen, protein CIP2A | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| p90 autoantigen (2G10) | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction