missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIP2A Antibody (2G10), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-59722MFV610
This item is not returnable.
View return policy
Description
CIP2A Monoclonal antibody specifically detects CIP2A in Human, Mouse, Rat, Primate samples. It is validated for Flow Cytometry
Specifications
| CIP2A | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate | |
| Purified |
| Flow Cytometry | |
| 2G10 | |
| Flow Cytometry | |
| Cancerous inhibitor of PP2A, CIP2A, FLJ12850, KIAA1524, MGC163436, p90, p90 autoantigen, protein CIP2A | |
| A synthetic peptide made to a C-terminal portion of the human CIP2A protein. [UniProt# Q8TCG1] | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 57650 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction