Learn More
CIP2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 11 publications
Brand:  Novus Biologicals NB100-68264
Product Code. 18788973
  • € 575.00 / 0.10mL
Description
CIP2A Polyclonal specifically detects CIP2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry, Immunohistochemistry (Paraffin)
RUO
IgG
Rabbit
Store at 4C. Do not freeze.