missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CIP2A Monoclonal Antibody (2G10), DyLight™ 488
GREENER_CHOICE

Product Code. 13267548
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13267548 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13267548 Supplier Invitrogen™ Supplier No. MA516082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CIP2A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Inhibition of protein phosphatase 2A (PP2A) activity has been identified as a prerequisite for the transformation of human cells. The protein, designated Cancerous Inhibitor of PP2A (CIP2A), interacts directly with the oncogenic transcription factor c-Myc, inhibits PP2A activity toward c-Myc serine 62 (S62), and thereby prevents c-Myc proteolytic degradation. In addition to its function in c-Myc stabilization, CIP2A promotes anchorage-independent cell growth and in vivo tumor formation. The oncogenic activity of CIP2A is demonstrated by transformation of human cells by overexpression of CIP2A. Importantly, CIP2A is overexpressed in two common human malignancies, head and neck squamous cell carcinoma (HNSCC) and colon cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIP2A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2G10
Concentration 1.1 mg/mL
Conjugate DyLight 488
Formulation 50mM sodium borate with 0.05% sodium azide
Gene Cip2a
Gene Accession No. Q8BWY9, Q8TCG1
Gene Alias AA408511; AU018569; C330027C09Rik; Cancerous inhibitor of PP2A; cancerous inhibitor of protein phosphatase 2A; Cip2a; Kiaa1524; p90; p90 autoantigen; p90 autoantigen homolog; Protein CIP2A; RGD1310335; RIKEN cDNA C330027C09 gene; similar to RIKEN cDNA C330027C09
Gene Symbols Cip2a
Host Species Mouse
Immunogen A synthetic peptide made to a C-terminal portion of the human CIP2A protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 224171, 360711, 57650
Target Species Human, Mouse, Monkey, Rat
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.