missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIP2A Monoclonal Antibody (2G10), DyLight™ 650

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA516081
This item is not returnable.
View return policy
Description
CIP2A Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Inhibition of protein phosphatase 2A (PP2A) activity has been identified as a prerequisite for the transformation of human cells. The protein, designated Cancerous Inhibitor of PP2A (CIP2A), interacts directly with the oncogenic transcription factor c-Myc, inhibits PP2A activity toward c-Myc serine 62 (S62), and thereby prevents c-Myc proteolytic degradation. In addition to its function in c-Myc stabilization, CIP2A promotes anchorage-independent cell growth and in vivo tumor formation. The oncogenic activity of CIP2A is demonstrated by transformation of human cells by overexpression of CIP2A. Importantly, CIP2A is overexpressed in two common human malignancies, head and neck squamous cell carcinoma (HNSCC) and colon cancer.
Specifications
| CIP2A | |
| Monoclonal | |
| 0.63 mg/mL | |
| 50mM sodium borate with 0.05% sodium azide | |
| Q8BWY9, Q8TCG1 | |
| Cip2a | |
| A synthetic peptide made to a C-terminal portion of the human CIP2A protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2G10 | |
| DyLight 650 | |
| Cip2a | |
| AA408511; AU018569; C330027C09Rik; Cancerous inhibitor of PP2A; cancerous inhibitor of protein phosphatase 2A; Cip2a; Kiaa1524; p90; p90 autoantigen; p90 autoantigen homolog; Protein CIP2A; RGD1310335; RIKEN cDNA C330027C09 gene; similar to RIKEN cDNA C330027C09 | |
| Mouse | |
| Protein G | |
| RUO | |
| 224171, 360711, 57650 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction