missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cip4 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-59833AF594
This item is not returnable.
View return policy
Description
Cip4 Polyclonal antibody specifically detects Cip4 in Human samples. It is validated for Western Blot
Specifications
| Cip4 | |
| Polyclonal | |
| Western Blot | |
| cdc42-interacting protein 4, CIP4STOT, HSTP, Protein Felic, Salt tolerant protein, salt tolerator, STPTRIP-10, thyroid hormone receptor interactor 10, thyroid receptor interacting protein 10, Thyroid receptor-interacting protein 10, TR-interacting protein 10 | |
| The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human thyroid receptor interacting protein 10 (Cdc42-interacting protein 4) using the numbering given in entry AAM46851.1 (GeneID 9322). | |
| 0.1 mL | |
| Cancer | |
| 9322 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction