missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ciprofloxacin-d8, TRC
Specifications
Specifications
| Chemical Name or Material | Ciprofloxacin-D8 (piperazine D8) |
| CAS | 1130050-35-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 2H8 H10 F N3 O3 |
| Quantity | 25 mg |
| Synonym | Ciprofloxacin-D8 (piperazine D8), 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl-2,2,3,3,5,5,6,6-d8)-, Ciprofloxacin-d8 |
| InChI Formula | InChI=1S/C17H18FN3O3/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24/h7-10,19H,1-6H2,(H,23,24)/i3D2,4D2,5D2,6D2 |
| SMILES | [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)C4CC4)C1([2H])[2H] |
| IUPAC Name | 1-cyclopropyl-6-fluoro-7-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid |
| Molecular Weight (g/mol) | 339.39 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?