Learn More
Ciprofloxacin Lactate, TRC
Specifications
Specifications
| Chemical Name or Material | Ciprofloxacin lactate |
| CAS | 97867-33-9 |
| Molecular Formula | C17 H18 F N3 O3 . C3 H6 O3 |
| Quantity | 100 mg |
| Synonym | 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-, mono(2-hydroxypropanoate) (9CI), Propanoic acid, 2-hydroxy-, compd. with 1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid (1:1) (9CI), Ciprofloxacin lactate |
| InChI Formula | InChI=1S/C17H18FN3O3.C3H6O3/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24;1-2(4)3(5)6/h7-10,19H,1-6H2,(H,23,24);2,4H,1H3,(H,5,6) |
| SMILES | CC(O)C(=O)O.OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(F)cc3C1=O |
| IUPAC Name | 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;2-hydroxypropanoic acid |
| Molecular Weight (g/mol) | 421.42 |
| Formula Weight | 421.1649 |