missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIRBP Antibody (SR1307), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22402-50ul
This item is not returnable.
View return policy
Description
CIRBP Monoclonal antibody specifically detects CIRBP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| CIRBP | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1307 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| A18 hnRNP, A18HNRNP, CIRPcold-inducible RNA-binding protein, cold inducible RNA binding protein, cold inducible RNA-binding protein, glycine-rich RNA binding protein, Glycine-rich RNA-binding protein CIRP | |
| A synthesized peptide derived from CIRBP (Uniprot #: Q14011) | |
| 50 μg | |
| Cell Biology, Cell Cycle and Replication | |
| 1153 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction