missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Circlegrow™ Agar

Product Code. 11367729 Shop All MP Biomedicals Products
Click to view available options
Quantity:
908 g
2.3 kg
Unit Size:
1kg
2.3kg

Product Code. 11367729

Brand: MP Biomedicals™ 113100132

Please to purchase this item. Need a web account? Register with us today!

Circlegrow™ Agar is designed for rapid growth of E. coli and high yields of plasmid DNA.

  • CIRCLEGROW™ Agar media is used for growth of E.coli.
  • It also extends plasmid production past log phase.
  • Acceptable pH range is 6.9 ± 0.3 at 25°C.
  • Please note that the pH of CIRCLEGROW™ Medium liquid increases as the temperature of the solution decreases.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 55 g/L
Form Powder
Grade Molecular Biology Certified™
Media Type Circlegrow
Preparation Method Reagents are tested and chosen for their ability to promote optimum growth of E. coli when combined in Circlegrow™ Agar formulations. Chosen reagents are combined in the accurate proportions according to the proprietary Circlegrow™ Agar formula and thoroughly blended for a uniform distribution.
Formulation Proprietary formulation containing Yeast Extract, Tryptone Broth, and Vitamins. Biological buffers have been added for proper pH adjustment.
For Use With (Application) Cell Culture, Proteomics, Purification and Detection, Recombinant Protein Expression and Analysis, Bacterial Culture, Plasmid Production, Protein Expression, Gene Expression, Cloning, Genetic Manipulation
Quantity 908 g
Solubility Soluble in water, 55 g/L
Synonym Enhanced Bacterial Media

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.