missing translation for 'onlineSavingsMsg'
Learn More

cis-1-(3-Chloroallyl)-3,5,7-triaza-1-azoniaadamantantane Chloride, TRC

Product Code. 30008868
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008868 1 g 1g
30138524 5 g 5g
30133900 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008868 Supplier Toronto Research Chemicals Supplier No. TRCC3642301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51229-78-8
Formula Weight 250.0752
InChI Formula InChI=1S/C9H16ClN4.ClH/c10-2-1-3-14-7-11-4-12(8-14)6-13(5-11)9-14;/h1-2H,3-9H2;1H/q+1;/p-1/b2-1-;
IUPAC Name 1-[(Z)-3-chloroprop-2-enyl]-3,5,7-triaza-1-azoniatricyclo[3.3.1.13,7]decane;chloride
Molecular Formula C9 H16 Cl N4 . Cl
Molecular Weight (g/mol) 251.16
SMILES [Cl-].Cl\C=C/C[N+]12CN3CN(CN(C3)C1)C2
Synonym 3,5,7-Triaza-1-azoniatricyclo[3.3.1.13,7]decane, 1-(3-chloro-2-propenyl)-, chloride, (Z)-, 3,5,7-Triaza-1-azoniatricyclo[3.3.1.13,7]decane, 1-[(2Z)-3-chloro-2-propenyl]-, chloride (9CI), 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, 3,5,7-triaza-1-azoniatricyclo[3.3.1.13,7]decane deriv., Dowicil 200, cis-1-(3-Chloroallyl)-3,5,7-triaza-1-azoniaadamantane chloride, cis-1-(3-Chloroallyl)-3,5,7-triaza-1-azoniaadamantantane chloride, cis-CTAC, Quaternium 15, 3,5,7-Triaza-1-azoniatricyclo[3.3.1.13,7]decane, 1-[(2Z)-3-chloro-2-propen-1-yl]-, chloride (1:1) (ACI)
Chemical Name or Material 1-cis-3-Chloroallyl-3,5,7-triaza-1-azonia-adamantane chloride

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.