Learn More
cis-2-Aminocyclohexanol hydrochloride, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 6936-47-6 |
| Molecular Formula | C6H14ClNO |
| MDL Number | MFCD00143981 |
| Quantity | 5 g |
| SMILES | Cl[H].N[C@H]1CCCC[C@H]1O |
| Molecular Weight (g/mol) | 151.63 |
| Percent Purity | 95% |
| Chemical Name or Material | cis-2-Aminocyclohexanol hydrochloride |