Learn More
cis-4-(Boc-amino)cyclohexaneacetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
cis-4-(Boc-amino)cyclohexaneacetic acid is generally used as a intermediate in chemical research.
Solubility
Slightly Soluble (1.1 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
cis-4-(Boc-amino)cyclohexaneacetic acid is generally used as a intermediate in chemical research.
Solubility
Slightly Soluble (1.1 g/L) (25°C).
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Solubility Information | Slightly Soluble (1.1g/L) (25°C). |
IUPAC Name | 2-[4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexyl]acetic acid |
Formula Weight | 257.32 |
Percent Purity | 97% |
Quantity | 250 mg |
Chemical Name or Material | cis-4-(Boc-amino)cyclohexaneacetic acid |