missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-4-Hydroxy-L-proline-2,5,5-d3, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | cis-4-Hydroxy-L-proline-d3 |
| Molecular Formula | C5 D3 H6 N O3 |
| Quantity | 0.05 g |
| Synonym | cis-4-Hydroxy-L-proline-2,5,5-d3, (4 S)-4-Hydroxy-L-proline-2,5,5-D3, cis-4-Hydroxy-L-proline-2,5,5-D3, (2 S,4 S)-4-Hydroxyproline-2,5,5-d3, (2 S,4 S)-4-Hydroxypyrrolidine-2,5,5-d3 acid |
| InChI Formula | InChI=1 S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7 H,1-2H2,(H,8,9)/t3-,4-/m0/s1/i2D2,4 D |
| SMILES | [2 H]C1([2 H])N[C@@]([2 H])(C[C@@H]1 O)C(=O)O |
| IUPAC Name | (2 S,4 S)-2,5,5-trideuterio-4-hydroxypyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 134.148 |
| Formula Weight | 134.077 g/mol |
| Percent Purity | 98 atom % D, also 15% deuterated (at positions 3 and 4), min 97% Chemical Purity) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction