Learn More
cis-5,8,11,14,17-Eicosapentaenoic acid, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 10417-94-4 |
| Molecular Formula | C20H30O2 |
| MDL Number | MFCD00065716 |
| Quantity | 1 g |
| SMILES | CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCC(O)=O |
| Molecular Weight (g/mol) | 302.45 |
| Percent Purity | 95% |
| Chemical Name or Material | cis-5,8,11,14,17-Eicosapentaenoic acid |
Product Title
Individuate un'opportunità di miglioramento?