Learn More
cis-5-Norbornene-endo-2,3-dicarboxylic anhydride, 97%
Product Code. 11481487
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481487 100 g 100g
11461487 5 g 5g
11471487 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11481487 Supplier Thermo Scientific Alfa Aesar Supplier No. B24951.22
Request Bulk or Custom Format
CAS: 129-64-6 | C9H8O3 | 164.16 g/mol
cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.
Solubility
It decomposes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 129-64-6
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00151106,MFCD00069013,MFCD00630580,MFCD00151106
InChI Key KNDQHSIWLOJIGP-UHFFFAOYNA-N
Synonym cis-5-norbornene-endo-2,3-dicarboxylic anhydride
PubChem CID 126963424
IUPAC Name 4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
SMILES O=C1OC(=O)C2C3CC(C=C3)C12
Specifications
Melting Point 164°C to 167°C
Density 1.417
Quantity 100 g
Beilstein 83657
Sensitivity Moisture sensitive
Merck Index 14,1796
Solubility Information It decomposes in water.
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material cis-5-Norbornene-endo-2,3-dicarboxylic anhydride