missing translation for 'onlineSavingsMsg'
Learn More

cis-beta-Asarone (>70%), TRC

Product Code. 30345978
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345978 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345978 Supplier Toronto Research Chemicals Supplier No. TRCA7802001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material beta-Asarone
CAS 5273-86-9
Molecular Formula C12 H16 O3
Quantity 1 g
Synonym Benzene, 1,2,4-trimethoxy-5-(1-propenyl)-, (Z)-, Benzene, 1,2,4-trimethoxy-5-(1Z)-1-propenyl- (9CI), Benzene, 1,2,4-trimethoxy-5-propenyl-, (Z)- (8CI), β-Asarone (6CI), 1,2,4-Trimethoxy-5-(1Z)-1-propen-1-ylbenzene, (Z)-Asarone, (Z)-Azarone, cis-Asarone, cis-Isoasarone, cis-β-Asarone
InChI Formula InChI=1S/C12H16O3/c1-5-6-9-7-11(14-3)12(15-4)8-10(9)13-2/h5-8H,1-4H3/b6-5-
SMILES COc1cc(OC)c(\C=C/C)cc1OC
IUPAC Name 1,2,4-trimethoxy-5-[(Z)-prop-1-enyl]benzene
Molecular Weight (g/mol) 208.25
Formula Weight 208.1099
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.