missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CISD1 Mouse anti-Human, Clone: OTI2B3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500904
This item is not returnable.
View return policy
Description
This gene encodes a protein with a CDGSH iron-sulfur domain and has been shown to bind a redox-active [2Fe-2S] cluster. The encoded protein has been localized to the outer membrane of mitochondria and is thought to play a role in regulation of oxidation. Genes encoding similar proteins are located on chromosomes 4 and 17, and a pseudogene of this gene is located on chromosome 2.Specifications
| CISD1 | |
| Monoclonal | |
| 0.72 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NZ45 | |
| CISD1 | |
| Full length human recombit protein of human CISD1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B3 | |
| Unconjugated | |
| CISD1 | |
| C10orf70, MDS029, ZCD1, mitoNEET | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55847 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction