missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CISD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97235-100ul
This item is not returnable.
View return policy
Description
CISD2 Polyclonal antibody specifically detects CISD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CISD2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| CDGSH iron sulfur domain 2, CDGSH iron-sulfur domain-containing protein 2, CDGSH2, Endoplasmic reticulum intermembrane small protein, ERISWFS2, Miner1Wolfram syndrome 2, mitoNEET related 1, MitoNEET-related 1 protein, NAF-1, Nutrient-deprivation autophagy factor-1, ZCD2CDGSH iron sulfur domain-containing protein 2, zinc finger, CDGSH-type domain 2 | |
| Produced in rabbits immunized with E. coli-derived Human CISD2 fragment. | |
| 100 μg | |
| Neuroscience, Signal Transduction | |
| 493856 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction