Learn More
CISD3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30480-1-AP-20UL
 View more versions of this product
Product Code. 18039495
  • € 145.00 / 20µL
Description
The CISD3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CISD3. This antibody recognizes human, mouse antigen. The CISD3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CISD3
450 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
CDGSH iron sulfur domain 3, CISD3, Miner2, MitoNEET related protein 2
20 μL
284106
Store at -20°C. Stable for one year after shipment.