missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cisplatin Modified DNA Antibody (CP9/19), Allophycocyanin/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50165APCCY7
This item is not returnable.
View return policy
Description
Cisplatin Modified DNA Monoclonal specifically detects Cisplatin Modified DNA in Non-species specific samples. It is validated for Flow Cytometry.
Specifications
| Cisplatin Modified DNA | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| Rat | |
| Protein G purified | |
| DNA Repair, Epigenetics | |
| CP9/19 recognises only the intra-strand cross-links formed by Cisplatin between adjacent purines. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| CP9/19 | |
| Flow Cytometry | |
| ICR4 | |
| Cisplatin modified native DNA | |
| 0.1 mL | |
| Primary | |
| Non-species specific | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction