missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cisplatin Modified DNA Antibody (CP9/19), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-50165CL1
This item is not returnable.
View return policy
Description
Cisplatin Modified DNA Monoclonal antibody specifically detects Cisplatin Modified DNA in Nonspecies specific samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin),Dot Blot
Specifications
| Cisplatin Modified DNA | |
| Monoclonal | |
| CoraFluor 1 | |
| ICR4 | |
| Cisplatin modified native DNA | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| CP9/19 | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction