Learn More
Cisplatin Modified DNA Antibody (CP9/19), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50165MFV610
This item is not returnable.
View return policy
Description
Cisplatin Modified DNA Monoclonal antibody specifically detects Cisplatin Modified DNA in Non-species specific samples. It is validated for Flow Cytometry
Specifications
| Flow Cytometry | |
| Flow Cytometry | |
| ICR4 | |
| Protein G purified | |
| Store at 4C in the dark. |
| Monoclonal | |
| 50mM Sodium Borate | |
| Rat | |
| RUO |