Learn More
Cisplatin Modified DNA Antibody (CP9/19), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50165MFV610
Product Code. 18705882
  • € 584.00 / 0.10mL
Description
Cisplatin Modified DNA Monoclonal antibody specifically detects Cisplatin Modified DNA in Non-species specific samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flow Cytometry
Flow Cytometry
ICR4
Protein G purified
Store at 4C in the dark.