missing translation for 'onlineSavingsMsg'
Learn More

Citadiol Hydrobromide, TRC

Product Code. 30136763
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136763 250 mg 250mg
17943465 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136763 Supplier Toronto Research Chemicals Supplier No. TRCC504800250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103146-26-5
Molecular Formula C20 H23 F N2 O2 . Br H
Molecular Weight (g/mol) 423.32
Synonym 4-[4-(dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl]-3-(hydroxymethyl)-Benzonitrile hydrobromide (1:1), Citadiol Hydrobromide, (4RS)-4-[4-Cyano-2-(hydroxymethyl)phenyl]-4-(4-fluorophenyl)-4-hydroxy-N,N-dimethylbutan-1-aminium Hydrobromide
IUPAC Name 4-[4-(dimethylamino)-1-(4-fluorophenyl)-1-hydroxybutyl]-3-(hydroxymethyl)benzonitrile;hydrobromide
SMILES Br.CN(C)CCCC(O)(c1ccc(F)cc1)c2ccc(cc2CO)C#N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H23FN2O2.BrH/c1-23(2)11-3-10-20(25,17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-24;/h4-9,12,24-25H,3,10-11,14H2,1-2H3;1H
Formula Weight 422.1005
Chemical Name or Material Citadiol Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.