missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED1 Antibody (OTI3C12), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71199JF646
This item is not returnable.
View return policy
Description
CITED1 Monoclonal specifically detects CITED1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CITED1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CITED1 | |
| Full length recombinant protein of human CITED1 (NP_004134) produced in E.coli. | |
| 0.1 mL | |
| 4435.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3C12 | |
| Western Blot, Immunohistochemistry | |
| cbp/p300-interacting transactivator 1, Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain1,MSG1Melanocyte-specific protein 1, melanocyte-specific gene 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction