missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED1 Monoclonal Antibody (OTI3C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807075
This item is not returnable.
View return policy
Description
This gene encodes a member of the CREB-binding protein/p300-interacting transactivator with Asp/Glu-rich C-terminal domain (CITED) family of proteins. The encoded protein, also known as melanocyte-specific gene 1, may function as a transcriptional coactivator and may play a role in pigmentation of melanocytes. Alternatively spliced transcript variants have been described.Specifications
| CITED1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99966 | |
| CITED1 | |
| Full length human recombit protein of human CITED1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C12 | |
| Unconjugated | |
| CITED1 | |
| MSG1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4435 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction