missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED4 Antibody (HT13-2D6.3), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-41572MFV500
This item is not returnable.
View return policy
Description
CITED4 Monoclonal antibody specifically detects CITED4 in Human samples. It is validated for Flow Cytometry
Specifications
| CITED4 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| HT13-2D6.3 | |
| Flow Cytometry | |
| cbp/p300-interacting transactivator 4, Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain4,MRG-2, MRG2, MSG1-related protein 2, transcriptional co-activator 4 | |
| N-terminal fusion protein to full-length human CITED4 [UniProt# Q96RK1] | |
| 0.1 mL | |
| Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators | |
| 163732 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction