Learn More
Invitrogen™ Citrate Synthase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596404
Description
Positive Samples: 22Rv1, Jurkat, 293T, HepG2, MCF7, HeLa, Mouse heart, Mouse liver, Rat testis; Cellular Location: Mitochondrion matrix Immunogen sequence: QLSAAVTALN SESNFARAYA QGISRTKYWE LIYEDSMDLI AKLPCVAAKI YRNLYREGSG IGAIDSNLDW SHNFTNMLGY TDHQFTELTR LYLTIHSDHE GGNVSAHTSH LVGSALSDPY LSFAAAMNGL AGPLHGLANQ EVLVWLTQLQ KEVGKDVSDE KLRDYIWNTL NSGRVVPGYG HAVLRKTDPR YTCQREFALK HLPNDPMFKL VAQLYKIVPN VLLEQGKAKN PWPNVDAHSG VLLQYYGMTE MNYYTVLFGV SRALGVLAQL IWSRALGFPL ERPKSMSTEG LMKFVDSKSG.
The protein encoded by this gene is a Krebs tricarboxylic acid cycle enzyme that catalyzes the synthesis of citrate from oxaloacetate and acetyl coenzyme A. The enzyme is found in nearly all cells capable of oxidative metabolism. This protein is nuclear encoded and transported into the mitochondrial matrix, where the mature form is found.
Specifications
| Citrate Synthase | |
| Polyclonal | |
| Unconjugated | |
| CS | |
| 2610511A05Rik; 9030605P22Rik; Ahl4; BB234005; Cis; citrate (Si)-synthase; citrate synthase; citrate synthase precursor (EC 4.1.3.7); citrate synthase, mitochondrial; citrate synthetase; Cs; wu:fb58e04; zgc:55507 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12974, 1431, 170587 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.03 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O75390, Q8VHF5, Q9CZU6 | |
| CS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 167-466 of human CS (NP_0040682). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.