missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Citrate Synthase Recombinant Rabbit Monoclonal Antibody (23GB780), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | Citrate Synthase |
|---|---|
| Clone | 23GB780 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088926
|
Invitrogen™
MA554212 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088266
|
Invitrogen™
MA554213 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Citrate Synthase Recombinant Monoclonal antibody specifically detects Citrate Synthase in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Citrate Synthase | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O75390, Q8VHF5, Q9CZU6 | |
| 12974, 1431, 170587 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB780 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2610511A05Rik; 9030605P22Rik; Ahl4; BB234005; Cis; citrate (Si)-synthase; citrate synthase; citrate synthase precursor (EC 4.1.3.7); citrate synthase, mitochondrial; citrate synthetase; Cs; wu:fb58e04; zgc:55507 | |
| A synthetic peptide of human CS (400-466AA). | |
| Primary | |
| -20°C | |
| CS |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title