missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citron Kinase Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28740AF350
This item is not returnable.
View return policy
Description
Citron Kinase Polyclonal antibody specifically detects Citron Kinase in Human samples. It is validated for Western Blot
Specifications
| Citron Kinase | |
| Polyclonal | |
| Western Blot | |
| CITK, citron (rho-interacting, serine/threonine kinase 21), Citron Rho-Interacting Kinase, Citron Rho-Interacting Serine/Threonine Kinase, CRIK, KIAA0949, MCPH17, Rho-interacting Serine/Threonine kinase 21, serine/threonine kinase 21, Serine/threonine-protein kinase 21, STK21 | |
| The immunogen recognized by this antibody maps to a region between residue 1925 and 1975 of human citron (rho-interacting, serine/threonine kinase 21) using the numbering given in entry NP_009105.1 (GeneID 11113). | |
| 0.1 mL | |
| Protein Kinase | |
| 11113 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction