Learn More
CK 1827452, TRC
Product Code. 30137988
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137988 10 mg 10mg
30005952 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137988 Supplier Toronto Research Chemicals Supplier No. TRCC54400010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 873697-71-3
Molecular Formula C20 H24 F N5 O3
Molecular Weight (g/mol) 401.43
Synonym 1-Piperazinecarboxylic acid, 4-[[2-fluoro-3-[[[(6-methyl-3-pyridinyl)amino]carbonyl]amino]phenyl]methyl]-, methyl ester (9CI, ACI), CK 1827452, Methyl 4-[[2-fluoro-3-[N'-(6-methylpyridin-3-yl)ureido]phenyl]methyl]piperazine-1-carboxylate, Omecamtiv mecarbil
IUPAC Name methyl 4-[[2-fluoro-3-[(6-methylpyridin-3-yl)carbamoylamino]phenyl]methyl]piperazine-1-carboxylate
SMILES COC(=O)N1CCN(Cc2cccc(NC(=O)Nc3ccc(C)nc3)c2F)CC1
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C20H24FN5O3/c1-14-6-7-16(12-22-14)23-19(27)24-17-5-3-4-15(18(17)21)13-25-8-10-26(11-9-25)20(28)29-2/h3-7,12H,8-11,13H2,1-2H3,(H2,23,24,27)
Formula Weight 401.1863
Chemical Name or Material CK 1827452