missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CK1 epsilon Polyclonal Antibody

Description
Immunogen sequence: EFSTYLNFCR SLRFDDKPDY SYLRQLFRNL FHRQGFSYDY VFDWNMLKFG AARNPEDVDR ERREHEREER MGQLRGSATR ALPPGPPTGA TANRLRSAAE PVASTPASRI QPAGNTSPRA ISRVDRERKV SMRLHRGAPA NVSSSDLTGR QEVSRIPASQ TSVPFDHLGK; Positive Samples: HeLa, 293T; Cellular Location: Cytoplasm, Nucleus.
CSNK1E (CK1 epsilon) is a serine/threonine kinase implicated in a wide range of cellular functions, including vesicular trafficking, DNA damage repair, cell cycle progression, and cytokinesis. CSNK1E phosphorylate key regulatory proteins involved in the control of cell differentiation, proliferation, chromosome segregation, circadian rhythms, and metabolic pathways. CK1 family members share a highly conserved kinase domain but differ in their variable N-and C-terminal domains. Mammals have seven family members: alpha, beta, gamma1, gamma2, gamma3, delta and epsilon. CKIe positively regulates Akt possibly by inhibiting PP2A. CKIe play an important role in neurodegenerative diseases.
Specifications
Specifications
| Antigen | CK1 epsilon |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.93 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CSNK1E |
| Gene Accession No. | P49674, Q9JMK2 |
| Gene Alias | AI426939; AI551861; AW457082; casein kinase 1; casein kinase 1 epsilon; casein kinase 1, epsilon; casein kinase I isoform epsilon; CK1e; CK1epsilon; CKI, epsilon; CKIe; CKIepsilon; CKI-epsilon; Csnk1e; HCKIE; KC1 epsilon; KC1epsilon; MGC10398; OTTHUMP00000063258; OTTHUMP00000063263; RP1-5O6.1; tau |
| Gene Symbols | CSNK1E |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?