missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CK1 gamma-2 Monoclonal Antibody (OTI4H3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526560
This item is not returnable.
View return policy
Description
CK1 gamma-2 Monoclonal Antibody for Western Blot, IHC (P)
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling.
Specifications
| CK1 gamma-2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P78368 | |
| Csnk1g2 | |
| Human recombinant protein fragment corresponding to amino acids 163-415 of CSNK1G2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H3 | |
| Unconjugated | |
| Csnk1g2 | |
| 2810429I12Rik; AI463719; casein kinase 1 gamma 2; casein kinase 1 gamma 2 isoform; casein kinase 1 isoform gamma-2; casein kinase 1, gamma 2; casein kinase I isoform gamma-2; CK Igamma2; CK1G2; CKI gamma 2; CKI gamma2; CKI-gamma 2; CKIgamma2; CK-Igamma2; CKI-gamma2; CKI-gamma-2; CKIgamma-2; Csnk1g2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1455 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction