missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CK1 gamma-3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582950
This item is not returnable.
View return policy
Description
Immunogen sequence: MFDYEYDWIG KQLPTPVGAV QQDPALSSNR EAHQHRDKMQ QSKNQVVSST NGELNTDDPT AGRSNAPITA PTEVEVMDET NCQKV Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 68%.
The casein kinase I (CKI) gene family encodes serine/threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric and range from 25 to 55 kD. They are ubiquitous, being found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells. In eukaryotic cells, CKI is in volved in regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.
Specifications
| CK1 gamma-3 | |
| Polyclonal | |
| Unconjugated | |
| CSNK1G3 | |
| 3300002K07Rik; C330049O21Rik; casein kinase 1 gamma 3; casein kinase 1, gamma 3; casein kinase I isoform gamma-3; CKI-gamma 3; CSNK1G3; CSNK1G3L | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1456 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6M4 | |
| CSNK1G3 | |
| Recombinant protein corresponding to Human CSNK1G3. Recombinant protein control fragment (Product #RP-91455). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction