missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKAP2 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-21390AF488
This item is not returnable.
View return policy
Description
CKAP2 Polyclonal antibody specifically detects CKAP2 in Human samples. It is validated for Western Blot
Specifications
| CKAP2 | |
| Polyclonal | |
| Western Blot | |
| CTCL tumor antigen se20-10, cytoskeleton associated protein 2, cytoskeleton-associated protein 2, LB1DKFZp686L1238, se20-10, TMAPFLJ10749, Tumor- and microtubule-associated protein, tumor-associated microtubule-associated protein | |
| The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human cytoskeleton associated protein 2 using the numbering given in entry NP_001091995.1 (GeneID 26586). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 26586 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction