Learn More
CKAP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25486-1-AP-150UL
 View more versions of this product
Product Code. 16874055
  • € 410.00 / 150µL
Description
Nedd4-binding partner-1 (N4BP1) is a developmentally expressed protein interactor and monoubiquitylation substrate of Nedd4, a HECT domain-containing ubiquitin ligase that mediates ubiquitylation and proteasome degradation of target proteins and may have a fundamental role to play in embryonic processes. N4BP1 can undergo Nedd4-mediated polyubiquitylation and proteosomal degradation. N4BP1 also functions as a negative regulator of AIF4 (also known as ITCH or Atrophin-1), an E3-ubiquitin ligase, by interacting with the second WW domain of AIF4 and impairing ubiquitination of substrates.
Specifications
Specifications
CKAP2
Polyclonal
Unconjugated
CKAP2
CKAP2, CTCL tumor antigen se20 10, LB1, se20 10, TMAP
Rabbit
Antigen Affinity Chromatography
RUO
26586
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WWK9
CKAP2
CKAP2 Fusion Protein Ag21901
150 μL