Learn More
CKAP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25486-1-AP-150UL
This item is not returnable.
View return policy
Description
Nedd4-binding partner-1 (N4BP1) is a developmentally expressed protein interactor and monoubiquitylation substrate of Nedd4, a HECT domain-containing ubiquitin ligase that mediates ubiquitylation and proteasome degradation of target proteins and may have a fundamental role to play in embryonic processes. N4BP1 can undergo Nedd4-mediated polyubiquitylation and proteosomal degradation. N4BP1 also functions as a negative regulator of AIF4 (also known as ITCH or Atrophin-1), an E3-ubiquitin ligase, by interacting with the second WW domain of AIF4 and impairing ubiquitination of substrates.Specifications
| CKAP2 | |
| Polyclonal | |
| Unconjugated | |
| CKAP2 | |
| CKAP2, CTCL tumor antigen se20 10, LB1, se20 10, TMAP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26586 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WWK9 | |
| CKAP2 | |
| CKAP2 Fusion Protein Ag21901 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |