missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKAP2L Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-21392V
This item is not returnable.
View return policy
Description
CKAP2L Polyclonal antibody specifically detects CKAP2L in Human samples. It is validated for Western Blot
Specifications
| CKAP2L | |
| Polyclonal | |
| Western Blot | |
| cytoskeleton associated protein 2-like, cytoskeleton-associated protein 2-like, FLJ40629, MGC39683 | |
| The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human cytoskeleton associated protein 2-like using the numbering given in entry NP_689728.3 (GeneID 150468). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 150468 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction