Learn More
Invitrogen™ CKAP5 Monoclonal Antibody (3C13)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA549287
Product Code. 17961011
  • € 426.00 / 100µg
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human U-87MG whole cell, human COLO-320 whole cell, human SW620 whole cell, human HEPG2 whole cell, human CACO-2 whole cell, rat brain tissue, mouse brain tissue. IHC: human liver cancer tissue, human rectal cancer tissue, human renal cancer tissue, rat brain tissue. ICC/IF: Hela cell. Flow: A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Colonic and hepatic tumor over-expressed protein (ch-TOG) belongs to the TOG/XMAP215/Dis1 evolutionarily ancient family of microtubule-associated proteins with isoforms found in all major kingdoms of eukaryotes. This protein is required for the assembly and function of the meiotic or mitotic spindles during cell division and is localized to microtubule-organizing centers (MTOCs) during interphase or to the spindle poles and centrosomes during mitosis or cytokinesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CKAP5
Monoclonal
500 μg/mL
PBS with 4mg trehalose and no preservative
A2AGT5, Q14008
CKAP5
E.coli-derived human ch TOG/CKAP5 recombinant protein (Position: M1-L221).
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
3C13
Unconjugated
CKAP5
3110043H24Rik; 4930432B04Rik; CHTOG; ch-TOG; ch-TOGp; CKAP5; colonic and hepatic tumor overexpressed gene protein; colonic and hepatic tumor over-expressed gene protein; colonic and hepatic tumor over-expressed protein; cytoskeleton associated protein 5; cytoskeleton-associated protein 5; D730027C18Rik; KIAA0097; mKIAA0097; MSPS; T25636; TOG; TOGp
Mouse
Antigen affinity chromatography
RUO
311191, 75786, 9793
-20°C
Lyophilized