missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKB Mouse anti-Human, Clone: 2B7C3, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | CKB |
|---|---|
| Clone | 2B7C3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808945
|
Proteintech
66764-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898935
|
Proteintech
66764-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.Specifications
| CKB | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B CK, CKB, CKBB, Creatine kinase B chain, Creatine kinase B type, creatine kinase, brain | |
| CKB | |
| IgG2a | |
| Protein A | |
| Antibody |
| 2B7C3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P12277 | |
| 1152 | |
| CKB Fusion Protein Ag7933 | |
| Primary | |
| -20°C | |
| CKB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title